5-Chloro-3-methyl-1,2,4-thiadiazole

Lieferant: Apollo Scientific
Achtung

OR52111-1G OR52111-250MG OR52111-5G
APOSOR52111-1GEA 108 EUR
APOSOR52111-1G APOSOR52111-250MG APOSOR52111-5G
5-Chloro-3-methyl-1,2,4-thiadiazole
5-Chloro-3-methyl-1,2,4-thiadiazole
Formel: C3H3ClN2S
Molecular Weight: 134,5873 g/mol
Siedepunkt: 94…107 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08234803
CAS-Nummer: 21734-85-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR