2-Methanesulphonylaminobenzoic acid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(Methylsulphonamido)benzoic acid

OR52170-1G OR52170-25G OR52170-5G
APOSOR52170-1GEA 23.2 EUR
APOSOR52170-1G APOSOR52170-25G APOSOR52170-5G
2-Methanesulphonylaminobenzoic acid 97%
2-Methanesulphonylaminobenzoic acid
Formel: C₈H₉NO₄S
Molecular Weight: 215,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00590092
CAS-Nummer: 162787-61-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR