3-Amino-2-chlor-6-methylpyridin 96%
Lieferant: Apollo Scientific
Synonyme:
3-Amino-2-chloro-6-picoline, 5-Amino-6-chloro-2-picoline
OR52200-25G
APOSOR52200-25GEA
108
EUR
APOSOR52200-25G
3-Amino-2-chlor-6-methylpyridin 96%
3-Amino-2-chloro-6-methylpyridine
Formel:
C₆H₇ClN₂ Molecular Weight: 142,5900 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03095213 CAS-Nummer: 39745-40-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...