3-Amino-6-chlor-4-methylpyridin 97%

Lieferant: Apollo Scientific

Synonyme: 3-Amino-6-chloro-4-picoline, 5-Amino-2-chloro-4-methylpyridine, 6-Chloro-4-methylpyridin-3-amine

OR52201-100G
APOSOR52201-100GEA 162 EUR
APOSOR52201-100G
3-Amino-6-chlor-4-methylpyridin 97%
3-Amino-6-chloro-4-methylpyridine
Formel: C₆H₇ClN₂
Molecular Weight: 142,59 g/mol
Schmelzpunkt: 69…73 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03427661
CAS-Nummer: 66909-38-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR