2-(Benzyloxy)benzaldehyd 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR52207-100G
OR52207-500G
APOSOR52207-100GEA
36.4
EUR
APOSOR52207-100G
APOSOR52207-500G
2-(Benzyloxy)benzaldehyd 98%
2-(Benzyloxy)benzaldehyd
Formel:
C₁₄H₁₂O₂ Molecular Weight: 212,25 g/mol Siedepunkt: 326 °C Dichte: 1,339 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016583 CAS-Nummer: 5896-17-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...