3-Nitro-4-chinolinol 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Hydroxy-3-nitroquinoline, 3-Nitroquinolin-4-ol
OR52247-25G
OR52247-100G
APOSOR52247-25GEA
77
EUR
APOSOR52247-25G
APOSOR52247-100G
3-Nitro-4-chinolinol 98%
3-Nitro-4-chinolinol
Formel:
C₉H₆N₂O₃ Molecular Weight: 190,1600 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00277722 CAS-Nummer: 50332-66-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...