4,8-Dichlorchinolin

Lieferant: Apollo Scientific

OR52250-10G OR52250-1G OR52250-5G
APOSOR52250-10GEA 444 EUR
APOSOR52250-10G APOSOR52250-1G APOSOR52250-5G
4,8-Dichlorchinolin
4,8-Dichloroquinoline
Formel: C₉H₅Cl₂N
Molecular Weight: 198,05 g/mol
Schmelzpunkt: 156…160 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00024008
CAS-Nummer: 21617-12-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR