1-(4-Bromophenyl)-2-methyl-1-propanone
Lieferant: Apollo Scientific
Synonyme:
1-(4-Bromophenyl)-2-methylpropan-1-one
OR52251-1G
OR52251-5G
APOSOR52251-1GEA
94.4
EUR
APOSOR52251-1G
APOSOR52251-5G
1-(4-Bromophenyl)-2-methyl-1-propanone
1-(4-Bromophenyl)-2-methyl-1-propanone
Formel:
C₁₀H₁₁BrO Molecular Weight: 227.10 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09891214 CAS-Nummer: 49660-93-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...