4-Bromo-2-chloropyrimidine 95+%
Lieferant: Apollo Scientific
|
Achtung
|
OR52309-1G
OR52309-500MG
OR52309-5G
APOSOR52309-1GEA
157
EUR
APOSOR52309-1G
APOSOR52309-500MG
APOSOR52309-5G
4-Bromo-2-chloropyrimidine 95+%
4-Bromo-2-chloropyrimidine
Formel:
C₄H₂BrClN₂ Molecular Weight: 193.428 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08272196 CAS-Nummer: 885702-34-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...