4-Amino-3-iod-1H-pyrazolo[3,4-d]pyrimidin 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-iodo-1H-pyrazolo[3,4-d]pyrimidin-4-amine
OR52321-1G
OR52321-25G
OR52321-5G
APOSOR52321-1GEA
52.2
EUR
APOSOR52321-1G
APOSOR52321-25G
APOSOR52321-5G
4-Amino-3-iod-1H-pyrazolo[3,4-d]pyrimidin 98%
4-Amino-3-iod-1H-pyrazolo[3,4-d]pyrimidin
Formel:
C₅H₄N₅I Molecular Weight: 261,03 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03787931 CAS-Nummer: 151266-23-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...