2-Methyl-m-anissäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Methoxy-2-methylbenzoic acid
OR52333-100G
OR52333-25G
OR52333-500G
APOSOR52333-100GEA
33.5
EUR
APOSOR52333-100G
APOSOR52333-25G
APOSOR52333-500G
2-Methyl-m-anissäure 98%
2-Methyl-m-anissäure
Formel:
C₉H₁₀O₃ Molecular Weight: 166,18 g/mol Schmelzpunkt: 149…151 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02094039 CAS-Nummer: 55289-06-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...