Tetrapropylammoniumperruthenat 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: TPAP, Tetra-n-propylammonium perruthenate(VII)

OR52342-1G OR52342-5G
APOSOR52342-1GEA 81.2 EUR
APOSOR52342-1G APOSOR52342-5G
Tetrapropylammoniumperruthenat 95%
Tetrapropylammonium perruthenate
Formel: C₁₂H₂₈NO₄Ru
Molecular Weight: 351,43 g/mol
Schmelzpunkt: 160 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00074914
CAS-Nummer: 114615-82-6
UN: 1479
ADR: 5.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR