IOX1 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 8-Hydroxyquinoline-5-carboxylic acid

OR52383-1G OR52383-100MG OR52383-250MG
APOSOR52383-1GEA 799 EUR
APOSOR52383-1G APOSOR52383-100MG APOSOR52383-250MG
IOX1 95%
IOX1
Formel: C₁₀H₇NO₃
Molecular Weight: 189,17 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD18417145
CAS-Nummer: 5852-78-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR