1-Ethynyl-3,5-dimethoxybenzene
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3,5-Dimethoxyphenylacetylene
OR52391-5G
APOSOR52391-5GEA
78.3
EUR
APOSOR52391-5G
1-Ethynyl-3,5-dimethoxybenzene
1-Ethynyl-3,5-dimethoxybenzene
Formel:
C₁₀H₁₀O₂ Molecular Weight: 162,19 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD03839985 CAS-Nummer: 171290-52-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...