Aminoguanidin Hydrochlorid

Lieferant: Apollo Scientific
Achtung

Synonyme: Aminoguanidine HCl, Aminoguanidine Hydrochloride, Guanylhydrazine hydrochloride

OR52394-100G OR52394-500G
APOSOR52394-100GEA 123 EUR
APOSOR52394-100G APOSOR52394-500G
Aminoguanidin Hydrochlorid
Aminoguanidin Hydrochlorid
Formel: CH₇ClN₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00039074
CAS-Nummer: 1937-19-5
EINECS: 217-707-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR