Methyl-5-hydroxy-2-methylbenzoate 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 5-hydroxy-2-methylbenzoate
OR52413-5G
OR52413-25G
APOSOR52413-5GEA
58
EUR
APOSOR52413-5G
APOSOR52413-25G
Methyl-5-hydroxy-2-methylbenzoate 95%
Methyl-5-hydroxy-2-methylbenzoate
Formel:
C₉H₁₀O₃ Molecular Weight: 166,18 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11226199 CAS-Nummer: 73505-48-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...