Fmoc-trans-4-(aminomethyl)cyclohexane-1-carboxylic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: Fmoc-trans-4-Amc-OH, trans-4-(((((9H-fluoren-9-yl)methoxy)carbonyl)amino)methyl)cyclohexanecarboxylic acid, N-Fmoc-tranexamic acid, trans-4-[[[[(9H-Fluoren-9-yl)methoxy]carbonyl]amino]methyl]cyclohexanecarboxylic acid, trans-4-(Fmoc-aminomethyl)cyclohexanecarboxylic acid

OR52526-25G
APOSOR52526-25GEA 109 EUR
APOSOR52526-25G
Fmoc-trans-4-(aminomethyl)cyclohexane-1-carboxylic acid
Fmoc-trans-4-(aminomethyl)cyclohexane-1-carboxylic acid
Formel: C₂₃H₂₅NO₄
Molecular Weight: 379.46 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00273480
CAS-Nummer: 167690-53-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR