1-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2,3,6-tetrahydropyridine 90%
Lieferant: Apollo Scientific
|
Achtung
|
OR52575-250MG
OR52575-1G
OR52575-500MG
APOSOR52575-250MGEA
403
EUR
APOSOR52575-250MG
APOSOR52575-1G
APOSOR52575-500MG
1-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2,3,6-tetrahydropyridine 90%
1-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2,3,6-tetrahydropyridine
Formel:
C₁₂H₂₂BNO₂ Lagertemperatur: Kühlschrank |
MDL:
MFCD16039627 CAS-Nummer: 1254982-25-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...