Boc-D-Ser(Me)-OH 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(R)-2-((tert-Butoxycarbonyl)amino)-3-methoxypropanoic acid, Boc-O-Methyl-D-serine
OR52578-25G
APOSOR52578-25GEA
81.2
EUR
APOSOR52578-25G
Boc-D-Ser(Me)-OH 97%
Boc-D-Ser(Me)-OH
Formel:
C₉H₁₇NO₅ Molecular Weight: 219,2 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08063987 CAS-Nummer: 86123-95-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...