Fmoc-Lys(Mtt)-OH 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Nα-[(9H-Fluoren-9-ylmethoxy)carbonyl]-Nepsilon-(4-methyltrityl)-L-lysine, Nα-Fmoc-Nepsilon-(4-methyltrityl)-L-lysine
OR52596-5G
OR52596-100G
OR52596-25G
APOSOR52596-5GEA
101
EUR
APOSOR52596-5G
APOSOR52596-100G
APOSOR52596-25G
Fmoc-Lys(Mtt)-OH 98%
Fmoc-Lys(Mtt)-OH
Formel:
C₄₁H₄₀N₂O₄ Molecular Weight: 624.781 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00237166 CAS-Nummer: 167393-62-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...