Fmoc-Glu-OtBu 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(S)-4-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-(tert-butoxy)-5-oxopentanoic acid, N-Fmoc-L-glutamic acid 1-tert-butyl ester, Fmoc-Glu-OtBu
OR52597-100G
OR52597-25G
APOSOR52597-100GEA
331
EUR
APOSOR52597-100G
APOSOR52597-25G
Fmoc-Glu-OtBu 98%
Fmoc-Glu-OtBu
Formel:
C₂₄H₂₇NO₆ Molecular Weight: 425,47 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00065648 CAS-Nummer: 84793-07-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...