Methylvanillat
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 4-hydroxy-3-methoxybenzoate
OR5261-100G
OR5261-25G
OR5261-500G
APOSOR5261-100GEA
65.4
EUR
APOSOR5261-100G
APOSOR5261-25G
APOSOR5261-500G
Methylvanillat
Methylvanillat
Formel:
C₉H₁₀O₄ Molecular Weight: 182,18 g/mol Siedepunkt: 285…287 °C Schmelzpunkt: 64…67 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008438 CAS-Nummer: 3943-74-6 EINECS: 223-525-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...