5-Amino-2-methylpyridin 98%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
6-Methylpyridin-3-amin, 3-Amino-6-picolin, 6-Methyl-3-pyridylamin, 5-Amino-2-picolin
OR5274-25G
OR5274-5G
APOSOR5274-25GEA
53.8
EUR
APOSOR5274-25G
APOSOR5274-5G
5-Amino-2-methylpyridin 98%
5-Amino-2-methylpyridin
Formel:
C₆H₈N₂ Molecular Weight: 108.14 g/mol Schmelzpunkt: 95…99 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00833389 CAS-Nummer: 3430-14-6 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...