2-Amino-3,4-dimethylbenzoesäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Amino-3,4-dimethylbenzoic Acid, ['3,4-Dimethylanthranilic acid']
OR5275-25G
OR5275-5G
APOSOR5275-25GEA
29
EUR
APOSOR5275-25G
APOSOR5275-5G
2-Amino-3,4-dimethylbenzoesäure
2-Amino-3,4-dimethylbenzoic acid
Formel:
C₉H₁₁NO₂ Molecular Weight: 165,19 g/mol Schmelzpunkt: 184…186 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00130041 CAS-Nummer: 50419-58-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...