1-(3-Bromphenyl)piperidin 96%

Lieferant: Apollo Scientific

OR52751-1G OR52751-5G
APOSOR52751-1GEA 42.2 EUR
APOSOR52751-1G APOSOR52751-5G
1-(3-Bromphenyl)piperidin 96%
1-(3-Bromphenyl)piperidin
Formel: C₁₁H₁₄BrN
Molecular Weight: 240,1396 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04108532
CAS-Nummer: 84964-24-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR