6-(1-Piperidinyl)-3-pyridinamine 95%

Lieferant: Apollo Scientific

OR52768-1G OR52768-250MG
APOSOR52768-1GEA 422 EUR
APOSOR52768-1G APOSOR52768-250MG
6-(1-Piperidinyl)-3-pyridinamine 95%
6-(1-Piperidinyl)-3-pyridinamine
Formel: C₁₀H₁₅N₃
Molecular Weight: 177.25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06447399
CAS-Nummer: 55403-29-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR