2-Phenylimidazo[1,2-a]pyridin-3-carbonsäure
Lieferant: Apollo Scientific
|
Achtung
|
OR5278-1G
OR5278-250MG
OR5278-100MG
APOSOR5278-1GEA
328
EUR
APOSOR5278-1G
APOSOR5278-250MG
APOSOR5278-100MG
2-Phenylimidazo[1,2-a]pyridin-3-carbonsäure
2-Phenylimidazo[1,2-a]pyridin-3-carbonsäure
Formel:
C₁₄H₁₀N₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00269226 CAS-Nummer: 123533-41-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...