3-Hydroxyanthranilsäure 98%

Lieferant: Apollo Scientific

Synonyme: 3-Hydroxyanthranilic acid

OR52785-10G OR52785-5G
APOSOR52785-10GEA 217 EUR
APOSOR52785-10G APOSOR52785-5G
3-Hydroxyanthranilsäure 98%
3-Hydroxyanthranilsäure
Formel: C₇H₇NO₃
Molecular Weight: 153,14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00007700
CAS-Nummer: 548-93-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR