3,8-Dichloro-1,5-naphthyridine 95%

Lieferant: Apollo Scientific

OR52789-1G OR52789-500MG
APOSOR52789-1GEA 957 EUR
APOSOR52789-1G APOSOR52789-500MG
3,8-Dichloro-1,5-naphthyridine 95%
3,8-Dichloro-1,5-naphthyridine
Formel: C₈H₄Cl₂N₂
Lagertemperatur: Kühlschrank
MDL: MFCD00034435
CAS-Nummer: 28252-81-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR