Dinatriumbenzol-1,3-disulfonat 94%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1,3-Benzenedisulphonic acid disodium salt, Benzene-1,3-disulphonic acid disodium salt

OR5283-100G OR5283-25G OR5283-500G
APOSOR5283-100GEA 29 EUR
APOSOR5283-100G APOSOR5283-25G APOSOR5283-500G
Dinatriumbenzol-1,3-disulfonat 94%
Dinatriumbenzol-1,3-disulfonat
Formel: C₆H₄Na₂O₆S₂
Molecular Weight: 282,21 g/mol
Schmelzpunkt: 300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007492
CAS-Nummer: 831-59-4
EINECS: 212-606-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR