4,6-Dichlorchinolin 98%

Lieferant: Apollo Scientific
Achtung

OR5287-1G OR5287-5G
APOSOR5287-1GEA 56.7 EUR
APOSOR5287-1G APOSOR5287-5G
4,6-Dichlorchinolin 98%
4,6-Dichloroquinoline
Formel: C₉H₅Cl₂N
Molecular Weight: 198,05 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00156141
CAS-Nummer: 4203-18-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR