Methyl-4-brom-2-nitrobenzoat 98%

Lieferant: Apollo Scientific

Synonyme: 4-Brom-2-nitrobenzoesäuremethylester

OR52909-100G OR52909-25G
APOSOR52909-100GEA 307 EUR
APOSOR52909-100G APOSOR52909-25G
Methyl-4-brom-2-nitrobenzoat 98%
Methyl-4-brom-2-nitrobenzoat
Formel: C₈H₆BrNO₄
Molecular Weight: 260,04 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10699645
CAS-Nummer: 158580-57-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR