1-Benzyl-1H-indazol-5-ylamine

Lieferant: Apollo Scientific
Achtung

OR53023-10G OR53023-1G OR53023-5G
APOSOR53023-10GEA 256 EUR
APOSOR53023-10G APOSOR53023-1G APOSOR53023-5G
1-Benzyl-1H-indazol-5-ylamine
1-Benzyl-1H-indazol-5-ylamine
Formel: C₁₄H₁₃N₃
Schmelzpunkt: 147…152 °C
Lagertemperatur: Kühlschrank
MDL: MFCD01719807
CAS-Nummer: 23856-21-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR