2,5-Dichlor-3-nitrobenzoesäure 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR53044-5G
APOSOR53044-5GEA
166
EUR
APOSOR53044-5G
2,5-Dichlor-3-nitrobenzoesäure 95%
2,5-Dichloro-3-nitrobenzoic acid
Formel:
C₇H₃Cl₂NO₄ Molecular Weight: 236,01 g/mol Schmelzpunkt: 216…220 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014691 CAS-Nummer: 88-86-8 EINECS: 201-862-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...