2-Amino-2-(4-methylphenyl)acetic acid 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Amino-2-(p-tolyl)acetic acid, H-DL-4-Me-Gly-OH
OR53059-1G
OR53059-5G
APOSOR53059-1GEA
72.5
EUR
APOSOR53059-1G
APOSOR53059-5G
2-Amino-2-(4-methylphenyl)acetic acid 98%
2-Amino-2-(4-methylphenyl)acetic acid
Formel:
C₉H₁₁NO₂ Molecular Weight: 165,19 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00665357 CAS-Nummer: 13227-01-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...