4-Amino-N,N-dimethylanilin 96%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
N,N-Dimethyl-1,4-phenylendiamin, N,N-Dimethyl-1,4-benzoldiamin, N,N-Dimethyl-p-phenylendiamin, N,N-Dimethyl-p-benzoldiamin
OR5307-250G
OR5307-50G
APOSOR5307-250GEA
325
EUR
APOSOR5307-250G
APOSOR5307-50G
4-Amino-N,N-dimethylanilin 96%
4-Amino-N,N-dimethylanilin
Redox indicator.
Formel:
C₈H₁₂N₂ Molecular Weight: 136,2 g/mol Siedepunkt: 262 °C Schmelzpunkt: 34…38 °C Flammpunkt: 130 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00007860 CAS-Nummer: 99-98-9 EINECS: 202-807-5 UN: 2811 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...