4'-Brom-3'-methylacetophenon

Lieferant: Apollo Scientific
Achtung

Synonyme: 4'-Bromo-3'-methoxyacetophenone

OR53079-1G OR53079-250MG
APOSOR53079-1GEA 87 EUR
APOSOR53079-1G APOSOR53079-250MG
4'-Brom-3'-methylacetophenon
4'-Brom-3'-methylacetophenon
Formel: C₉H₉BrO
Molecular Weight: 229,0706 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17169058
CAS-Nummer: 50870-44-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR