Boc-S-trityl-L-homocysteine

Lieferant: Apollo Scientific
Achtung

Synonyme: (S)-2-((tert-Butoxycarbonyl)amino)-4-(tritylthio)butanoic acid, Boc-Homocys(Trt)-OH, Boc-HoCys(Trt)-OH

OR53086-1G OR53086-250MG
APOSOR53086-1GEA 66.7 EUR
APOSOR53086-1G APOSOR53086-250MG
Boc-S-trityl-L-homocysteine
Boc-S-trityl-L-homocysteine
Formel: C₂₈H₃₁NO₄S
Molecular Weight: 477.619 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00672309
CAS-Nummer: 201419-16-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR