4-Nitro-o-phenylendiamin

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Nitro-1,2-phenylenediamine, 4-Nitrobenzene-1,2-diamine

OR53090-100G OR53090-25G
APOSOR53090-100GEA 45.1 EUR
APOSOR53090-100G APOSOR53090-25G
4-Nitro-o-phenylendiamin
4-Nitro-o-phenylendiamin
Formel: C₆H₇N₃O₂
Molecular Weight: 153,14 g/mol
Schmelzpunkt: 199…201 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007724
CAS-Nummer: 99-56-9
EINECS: 202-766-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR