5-Nitrobenzimidazol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Nitro-1H-benzimidazole
OR53095-100G
OR53095-25G
APOSOR53095-100GEA
132
EUR
APOSOR53095-100G
APOSOR53095-25G
5-Nitrobenzimidazol
5-Nitrobenzimidazole
Formel:
C₇H₅N₃O₂ Molecular Weight: 163.14 g/mol Schmelzpunkt: 207…211 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005604 CAS-Nummer: 94-52-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...