3-(tert-Butoxycarbonyl)-1,3-thiazolane-2-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1,3-Thiazolidine-2-carboxylic acid, N-BOC protected
OR53118-1G
OR53118-5G
APOSOR53118-1GEA
234
EUR
APOSOR53118-1G
APOSOR53118-5G
3-(tert-Butoxycarbonyl)-1,3-thiazolane-2-carboxylic acid
3-(tert-Butoxycarbonyl)-1,3-thiazolane-2-carboxylic acid
Formel:
C₉H₁₅NO₄S Molecular Weight: 233,29 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01569306 CAS-Nummer: 141783-63-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...