Ethyl-5-amino-1,3,4-oxadiazole-2-carboxylate 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Ethyl 5-amino-1,3,4-oxadiazole-2-carboxylate
OR53168-1G
OR53168-5G
APOSOR53168-1GEA
21.9
EUR
APOSOR53168-1G
APOSOR53168-5G
Ethyl-5-amino-1,3,4-oxadiazole-2-carboxylate 98%
Ethyl-5-amino-1,3,4-oxadiazole-2-carboxylate
Formel:
C₅H₇N₃O₃ Molecular Weight: 157,13 g/mol Schmelzpunkt: 200…205 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD03425200 CAS-Nummer: 4970-53-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...