3-(Dimethylphosphono)-N-methylolpropionamide
Lieferant: Apollo Scientific
|
Achtung
|
OR53172-100G
APOSOR53172-100GEA
314
EUR
APOSOR53172-100G
3-(Dimethylphosphono)-N-methylolpropionamide
3-(Dimethylphosphono)-N-methylolpropionamide
Formel:
C₆H₁₄NO₅P Lagertemperatur: Raumtemperatur |
MDL:
MFCD00468093 CAS-Nummer: 20120-33-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...