Ethyl-4-chloro-6-methyl-2-(methylthio)pyrimidine-5-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Ethyl 4-chloro-6-methyl-2-(methylthio)pyrimidine-5-carboxylate, Ethyl 4-chloro-6-methyl-2-(methylsulphanyl)pyrimidine-5-carboxylate
OR53212-10G
OR53212-1G
OR53212-5G
APOSOR53212-10GEA
996
EUR
APOSOR53212-10G
APOSOR53212-1G
APOSOR53212-5G
Ethyl-4-chloro-6-methyl-2-(methylthio)pyrimidine-5-carboxylate
Ethyl-4-chloro-6-methyl-2-(methylthio)pyrimidine-5-carboxylate
Formel:
C₉H₁₁ClN₂O₂S Lagertemperatur: Kühlschrank |
MDL:
MFCD08063161 CAS-Nummer: 583878-42-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...