5-Chloro-2,3-dihydro-(1H)-indole 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Chloroindoline

OR5335-1G OR5335-25G OR5335-5G
APOSOR5335-1GEA 79.9 EUR
APOSOR5335-1G APOSOR5335-25G APOSOR5335-5G
5-Chloro-2,3-dihydro-(1H)-indole 97%
5-Chloro-2,3-dihydro-(1H)-indole
Formel: C₈H₈ClN
Molecular Weight: 153,61 g/mol
Siedepunkt: 93…98 °C/0,4mmHg
Lagertemperatur: Gefrierschrank
MDL: MFCD01941330
CAS-Nummer: 25658-80-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR