4-Pentylbenzolsulfonylchlorid

Lieferant: Apollo Scientific
Gefahr

Synonyme: 4-Amylbenzol-1-sulfonylchlorid, 4-Pentylbenzolsulfonsäurechlorid, 4-Pentylbenzolsulfochlorid, 4-Amylbenzolsulfochlorid, 4-Pentylbenzol-1-sulfonylchlorid, 4-Amylbenzolsulfonsäurechlorid, 4-Amylbenzolsulfonylchlorid

OR5342-1G OR5342-25G OR5342-5G
APOSOR5342-1GEA 21.9 EUR
APOSOR5342-1G APOSOR5342-25G APOSOR5342-5G
4-Pentylbenzolsulfonylchlorid
4-Pentylbenzolsulfonylchlorid
Formel: C₁₁H₁₅ClO₂S
Molecular Weight: 246,76 g/mol
Schmelzpunkt: 39…44 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00173759
CAS-Nummer: 73948-18-2
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR