3,5-Dichlorpyridin 98%

Lieferant: Apollo Scientific
Achtung

OR5363-100G OR5363-25G
APOSOR5363-100GEA 48 EUR
APOSOR5363-100G APOSOR5363-25G
3,5-Dichlorpyridin 98%
3,5-Dichloropyridine
Formel: C₅H₃Cl₂N
Molecular Weight: 147,99 g/mol
Schmelzpunkt: 65…67 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006376
CAS-Nummer: 2457-47-8
EINECS: 219-537-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR