(Phenylsulphonyl)acetic acid 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR5366-1G
OR5366-25G
OR5366-5G
APOSOR5366-1GEA
65.5
EUR
APOSOR5366-1G
APOSOR5366-25G
APOSOR5366-5G
(Phenylsulphonyl)acetic acid 98%
(Phenylsulphonyl)acetic acid
Formel:
C₈H₈O₄S Schmelzpunkt: 109…114 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00025042 CAS-Nummer: 3959-23-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...