1-Boc-piperidin-4-carbonsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(tert-Butoxycarbonyl)piperidine-4-carboxylic acid, N-BOC-4-Piperidinecarboxylic acid, Boc-Inp-OH, 1-Boc-isonipecotic acid
OR5410-25G
OR5410-100G
APOSOR5410-25GEA
23.2
EUR
APOSOR5410-25G
APOSOR5410-100G
1-Boc-piperidin-4-carbonsäure 98%
1-Boc-piperidin-4-carbonsäure
Formel:
C₁₁H₁₉NO₄ Molecular Weight: 229.3 g/mol Schmelzpunkt: 148 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00076999 CAS-Nummer: 84358-13-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...