N-(4-Aminophenyl)Maleimid

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(4-Aminophenyl)-1H-pyrrole-2,5-dione

OR54116-1G OR54116-5G OR54116-250MG
APOSOR54116-1GEA 277 EUR
APOSOR54116-1G APOSOR54116-5G APOSOR54116-250MG
N-(4-Aminophenyl)Maleimid
N-(4-Aminophenyl)Maleimid
Formel: C₁₀H₈N₂O₂
Molecular Weight: 188,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00051191
CAS-Nummer: 29753-26-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR